missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DiscoverX PathHunter™ eXpress PRLR-JAK2 Functional Assay
missing translation for 'orderingAttributeHoverText'
Quantity:
1,000 dp (10 x 96-well)
missing translation for 'unitSize'
1 set
Description
The PathHunter™ PRLR-JAK2 Functional assay eXpress kit provides an easy-to-use cell based assay to study drug candidates (small molecules or biologics) targeting CTKs. This kit assesses PRLR activation via detection of JAK2 mediated phosphorylation-induced SH2 domain protein recruitment. This product uses EFC technology. The included cells overexpress PK-tagged PRLR cytokine receptor, an untagged CTK JAK2 and EA-tagged SH2 domain protein. Upon activation of the cytokine receptor, CTK-based receptor phosphorylation leads to SH2-EA recruitment and production of EFC signal. eXpress kits are a convenient, ready-to-use format which contains all materials needed to run the assay, including single-use vials of cryopreserved cells, cell plating media, detection reagent, and assay plates.
Specifications
Specifications
| Assay Entry | Kinase Activity |
| For Use With (Application) | Assay-Ready eXpress Kits |
| Quantity | 1,000 dp (10 x 96-well) |
| Target Entry | PRLR-JAK2 |
| Target | PRLR-JAK2 |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction