missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DiscoverX PathHunter™ eXpress GPR4 CHO-K1 β-Arrestin Orphan GPCR Assay
Description
The PathHunter™ GPR4 eXpress kit provides an easy-to-use cell based assay to study drug candidates (small molecules or biologics) targeting GPCRs. This kit assesses ligand based activation of GPR4 GPCR activity via detection of β-arrestin recruitment. This product uses EFC technology (www.discoverx.com/technologies-platforms/enzyme-fragment-complementation-technology). The included cells overexpress PK-tagged GPR4 and EA-tagged β-Arrestin 2. Activation of GPR4 stimulates the recruitment of β-Arrestin 2 and produces EFC signal. eXpress kits are a convenient, ready-to-use format which contains all materials needed to run the assay, including single-use vials of cryopreserved cells, cell plating media, detection reagent, and assay plates. Orphan receptor with unknown ligand.
Specifications
Specifications
| Assay Entry | GPCR |
| For Use With (Application) | Assay-Ready eXpress Kits |
| Quantity | 200 dp (2 x 96-well) |
| Target Entry | GPR4 |
| Cell Line | CHO-K1 |
| Target | GPR4 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?