missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DiscoverX PathHunter™ eXpress GPR18 CHO-K1 β-Arrestin Orphan GPCR Assay
Click to view available options
Quantity:
1,000 dp (10 x 96-well)
Unit Size:
1 set
Description
The PathHunter™ GPR18 eXpress kit provides an easy-to-use cell based assay to study drug candidates (small molecules or biologics) targeting GPCRs. This kit assesses ligand based activation of GPR18 GPCR activity via detection of β-arrestin recruitment. This product uses EFC technology (www.discoverx.com/technologies-platforms/enzyme-fragment-complementation-technology). The included cells overexpress PK-tagged GPR18 and EA-tagged β-Arrestin 2. Activation of GPR18 stimulates the recruitment of β-Arrestin 2 and produces EFC signal. eXpress kits are a convenient, ready-to-use format which contains all materials needed to run the assay, including single-use vials of cryopreserved cells, cell plating media, detection reagent, and assay plates. Orphan receptor with unknown ligand.
Specifications
Specifications
| Assay Entry | GPCR |
| For Use With (Application) | Assay-Ready eXpress Kits |
| Quantity | 1,000 dp (10 x 96-well) |
| Target Entry | GPR18 |
| Cell Line | CHO-K1 |
| Target | GPR18 |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction