missing translation for 'onlineSavingsMsg'
Learn More

DiscoverX PathHunter™ eXpress GLP1R Activated GPCR Internalization Assay

Product Code. 17363719
Encompass
Change view
Click to view available options
Quantity:
200 dp (2 x 96-well)
Unit Size:
1 set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17363719 200 dp (2 x 96-well) 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17363719 Supplier DiscoverX Supplier No. 930724E3CP0M

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

The PathHunter™ eXpress GLP1R Activated GPCR Internalization Assay measures GLP1R (GPCR) internalization via early-endosome localization. eXpress kits contain all assay materials: cells, reagents, and plates.

The PathHunter™ GLP1R Activated GPCR internalization eXpress kit provides an easy to-use cell based assay to study drug candidates (small molecules or biologics) affecting GPCR activation and internalization. This kit assesses ligand (e.g. exendin-4) based internalization of GLP1R via detection of localization to the early endosome. This product uses EFC technology. The included cells overexpress EA-tagged β-arrestin 2 as well as a PK-tagged early endosome protein and untagged GLP1R. Upon ligand-induced receptor activation, β-arrestin 2 is recruited to the activated GPCR and the resulting GLP1R-arrestin complex is internalized to the early endosome, leading to production of EFC signal. eXpress kits are a convenient, ready-to-use format which contains all materials needed to run the assay, including single-use vials of cryopreserved cells, cell plating media, detection reagent, and assay plates.

Specifications

Assay Entry GPCR
For Use With (Application) Assay-Ready eXpress Kits
Quantity 200 dp (2 x 96-well)
Target Entry GLP1R
Target GLP1R
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.