missing translation for 'onlineSavingsMsg'
Learn More

DiscoverX PathHunter™ eXpress FPR1 Total GPCR Internalization Assay

Product Code. 17373929
Encompass
Change view
Click to view available options
Quantity:
200 dp (2 x 96-well)
Unit Size:
1 set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17373929 200 dp (2 x 96-well) 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17373929 Supplier DiscoverX Supplier No. 930873E3CP0M

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

The PathHunter™ eXpress FPR1 Total GPCR Internalization Assay measures FPR1 (GPCR) internalization via early-endosome localization. eXpress kits contain all assay materials: cells, reagents, and plates.

The PathHunter™ FPR1 Total GPCR internalization eXpress kit provides an easy to-use cell based assay to study drug candidates (small molecules or biologics) affecting GPCR activation and internalization. This kit assesses ligand (e.g. formylpeptides) based internalization of FPR1 via detection of localization to the early endosome. This product uses EFC technology. The included cells overexpress PK-tagged FPR1 and an EA-tagged early endosome protein. Upon ligand-induced receptor activation, resulting internalization of the FPR1 receptor to the early endosome leads to production of EFC signal. eXpress kits are a convenient, ready-to-use format which contains all materials needed to run the assay, including single-use vials of cryopreserved cells, cell plating media, detection reagent, and assay plates.

Specifications

Assay Entry Total Internalization
For Use With (Application) Assay-Ready eXpress Kits
Primary Agonist WKYMVm-NH2
Quantity 200 dp (2 x 96-well)
Target Entry FPR1
Cell Line U2OS
Target FPR1
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.