missing translation for 'onlineSavingsMsg'
Learn More

DiscoverX PathHunter™ eXpress FPR1 Total GPCR Internalization Assay

Product Code. 17363929
Encompass
Change view
Click to view available options
Quantity:
1,000 dp (10 x 96-well)
Unit Size:
1 set
Product Code. Quantity unitSize
17363929 1,000 dp (10 x 96-well) 1 set
1 options
This item is not returnable. View return policy

Product Code. 17363929

Brand: DiscoverX 930873E3CP0L

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

The PathHunter™ eXpress FPR1 Total GPCR Internalization Assay measures FPR1 (GPCR) internalization via early-endosome localization. eXpress kits contain all assay materials: cells, reagents, and plates.

The PathHunter™ FPR1 Total GPCR internalization eXpress kit provides an easy to-use cell based assay to study drug candidates (small molecules or biologics) affecting GPCR activation and internalization. This kit assesses ligand (e.g. formylpeptides) based internalization of FPR1 via detection of localization to the early endosome. This product uses EFC technology. The included cells overexpress PK-tagged FPR1 and an EA-tagged early endosome protein. Upon ligand-induced receptor activation, resulting internalization of the FPR1 receptor to the early endosome leads to production of EFC signal. eXpress kits are a convenient, ready-to-use format which contains all materials needed to run the assay, including single-use vials of cryopreserved cells, cell plating media, detection reagent, and assay plates.

Specifications

Assay Entry Total Internalization
For Use With (Application) Assay-Ready eXpress Kits
Primary Agonist WKYMVm-NH2
Quantity 1,000 dp (10 x 96-well)
Target Entry FPR1
Cell Line U2OS
Target FPR1
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.