missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DiscoverX PathHunter™ eXpress cINSRa Functional Assay
Description
The PathHunter™ eXpress cINSRa Functional Assay provides an easy-to-use cell based assay to study drug candidates (small molecules or biologics) targeting RTKs. This kit assesses RTK activation via detection of activated cINSRa dimerization-induced SH2 domain protein recruitment. This product uses EFC technology. The included cells overexpress PK-tagged cINSRa and an EA-tagged SH2 domain protein. Upon ligand-induced receptor activation, SH2-EA recruitment leads to production of EFC signal. eXpress kits are a convenient, ready-to-use format which contains all materials needed to run the assay, including single-use vials of cryopreserved cells, cell plating media, detection reagent, and assay plates. Please note that this product expresses the Cyno version of the protein.
Specifications
Specifications
| Assay Entry | Receptor Phosphorylation |
| For Use With (Application) | Assay-Ready eXpress Kits |
| Primary Agonist | Insulin, Human |
| Quantity | 1,000 dp (10 x 96-well) |
| Target Entry | cINSRa |
| Cell Line | U2OS |
| Target | cINSRa |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?