missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DiscoverX PathHunter™ eXpress CDC25A Degradation Assay
Click to view available options
Quantity:
200 dp (2 x 96-well)
Unit Size:
1 set
Description
The PathHunter™ eXpress CDC25A Degradation Assay provides an easy-to-use cell based assay to study drug candidates (small molecules or biologics) targeting the cell cycle checkpoint pathway. This kit enables detection of levels of CDC25a, allowing for assessment of CHK-1-activation-induced CDC25a degradation. This product uses EFC technology. The cell line included overexpresses CDC25a target protein fused to PL. EA is contained in the detection reagent. Upon pathway-activation-induced degradation of CDC25a EFC signal is lost. eXpress kits are a convenient, ready-to-use format which contains all materials needed to run the assay, including single-use vials of cryopreserved cells, cell plating media, detection reagent, and assay plates.
Specifications
Specifications
| Assay Entry | Pathway |
| For Use With (Application) | Assay-Ready eXpress Kits |
| Format | 200 dp (2 x 96-well) |
| Quantity | 200 dp (2 x 96-well) |
| Target Entry | CDC25A |
| Target | CDC25A |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction