missing translation for 'onlineSavingsMsg'
Learn More

DiscoverX PathHunter™ eXpress CCR6 Activated GPCR Internalization Assay

Product Code. 17334119
Encompass
Click to view available options
Quantity:
1,000 dp (10 x 96-well)
Unit Size:
1 set
This item is not returnable. View return policy

Product Code. 17334119

Brand: DiscoverX 930966E3CP5L

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

The PathHunter™ eXpress CCR6 Activated GPCR Internalization Assay measures CCR6 (GPCR) internalization via early-endosome localization. eXpress kits contain all assay materials: cells, reagents, and plates.

The PathHunter™ CCR6 Activated GPCR internalization eXpress kit provides an easy to-use cell based assay to study drug candidates (small molecules or biologics) affecting GPCR activation and internalization. This kit assesses ligand (e.g. CCL20) based internalization of CCR6 via detection of localization to the early endosome. This product uses EFC technology. The included cells overexpress EA-tagged β-arrestin 2 as well as a PK-tagged early endosome protein and untagged CCR6. Upon ligand-induced receptor activation, β-arrestin 2 is recruited to the activated GPCR and the resulting CCR6-arrestin complex is internalized to the early endosome, leading to production of EFC signal. eXpress kits are a convenient, ready-to-use format which contains all materials needed to run the assay, including single-use vials of cryopreserved cells, cell plating media, detection reagent, and assay plates.

Specifications

Assay Entry β-Arrestin-2 recruitment
For Use With (Application) Assay-Ready eXpress Kits
Primary Agonist Recombinant Human MIP-3α (CCL20)
Quantity 1,000 dp (10 x 96-well)
Target Entry CCR6
Cell Line U2OS
Target CCR6
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.