missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DiscoverX PathHunter™ CHO-K1 MRGPRX2 Bioassay Kit
Click to view available options
Quantity:
1,000 dp (10 x 96-well)
Unit Size:
1 set
Description
The PathHunter™ CHO-K1 MRGPRX2 Bioassay Kit provides an easy-to-use cell based assay to measure drug potency and detect neutralizing antibodies. This bioassay asseses ligand (e.g. Cortistatin 14) based on activation of MRGPRX2 GPCR activity via detection of β-Arrestin recruitment. The included cell line overexpresses ProLink-tagged MRGPRX2 and EA-tagged . Activation of MRGPRX2 stimulates the recruitment of and produces EFC signal. Bioassay kits are a convenient, ready-to-use format which contains all materials needed to run the assay, including single-use vials of cryopreserved cells, cell plating media, control agonist, detection reagent, and assay plates.
Specifications
Specifications
| Assay Entry | β-Arrestin recruitment |
| For Use With (Application) | Assay-Ready Bioassay Kits |
| Quantity | 1,000 dp (10 x 96-well) |
| Cell Line | CHO-K1 |
| Target | MRGPRX2 |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction