missing translation for 'onlineSavingsMsg'
Learn More

Parvovirus NS1 Antibody (8E6E10) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody

£163.00 - £437.00

Specifications

Clone 8E6E10
Concentration 1 mg/mL
Dilution Western Blot 1:500-1:2000, Flow Cytometry 1:200-1:400, ELISA 1:10000, Immunocytochemistry/Immunofluorescence 1:200-1:1000
Applications Western Blot, Flow Cytometry, ELISA, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18001196
View Documents
Novus Biologicals
NBP2-61903
0.1 mL
£437.00
0.10mL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18673557
View Documents
Novus Biologicals
NBP2-61903-0.025ml
0.025 mL
£163.00
0.03mL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Parvovirus NS1 Monoclonal specifically detects Parvovirus NS1 in Human, Viral samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence.
Specifications

Specifications

8E6E10
Western Blot 1:500-1:2000, Flow Cytometry 1:200-1:400, ELISA 1:10000, Immunocytochemistry/Immunofluorescence 1:200-1:1000
Monoclonal
Purified
Human, Virus
Purified recombinant fragment of human Parvovirus NS1 (AA: 1-216) expressed in E. Coli.
Primary
585 kDa
1 mg/mL
Western Blot, Flow Cytometry, ELISA, Immunocytochemistry, Immunofluorescence
Unconjugated
Mouse
NS1
IgG1
Protein G purified
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.