missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Parvovirus NS1 Antibody (8E6E10) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-61903
This item is not returnable.
View return policy
Description
Parvovirus NS1 Monoclonal specifically detects Parvovirus NS1 in Human, Viral samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence.
Specifications
| Parvovirus NS1 | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:500-1:2000, Flow Cytometry 1:200-1:400, ELISA 1:10000, Immunocytochemistry/Immunofluorescence 1:200-1:1000 | |
| NS1 | |
| Purified recombinant fragment of human Parvovirus NS1 (AA: 1-216) expressed in E. Coli. | |
| Protein G purified | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG1 |
| Western Blot, Flow Cytometry, ELISA, Immunocytochemistry, Immunofluorescence | |
| 8E6E10 | |
| Unconjugated | |
| PBS with 0.05% Sodium Azide | |
| Mouse | |
| 585 kDa | |
| 0.1 mL | |
| Human, Virus | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction