missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Parthenolide

Product Code. 15812464 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
100 mg
500 mg
Unit Size:
100mg
500mg
This item is not returnable. View return policy

Product Code. 15812464

Brand: Thermo Scientific Chemicals 462151000

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 20554-84-1 | C15H20O3 | 248.32 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 20554-84-1
Molecular Formula C15H20O3
Molecular Weight (g/mol) 248.32
InChI Key KTEXNACQROZXEV-QLIGOWBFSA-N
IUPAC Name (1S,2S,4R,7Z,11S)-4,8-dimethyl-12-methylidene-3,14-dioxatricyclo[9.3.0.0²,⁴]tetradec-7-en-13-one
SMILES C\C1=C\CC[C@@]2(C)O[C@H]2[C@H]2OC(=O)C(=C)[C@@H]2CC1

Specifications

Melting Point 115.0°C
Color Clear
Quantity 100 mg
Merck Index 15, 7152
Solubility Information Solubility aq. soln.: .. Other solubilities: soluble in chloroform
Formula Weight 248.32
Chemical Name or Material Parthenolide

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.