CAS RN 20554-84-1
CAS RN 20554-84-1
IUPAC Name:
(1S,2S,4R,7Z,11S)-4,8-dimethyl-12-methylidene-3,14-dioxatricyclo[9.3.0.0²,⁴]tetradec-7-en-13-one
Synonyms:
parthenolide
Molecular Weight (g/mol):
248.32
Molecular Formula:
C15H20O3
InChi Key:
KTEXNACQROZXEV-QLIGOWBFSA-N
SMILES:
C\C1=C\CC[C@@]2(C)O[C@H]2[C@H]2OC(=O)C(=C)[C@@H]2CC1
1
–
2
of
2
results
1
Thermo Scientific Chemicals Parthenolide
CAS: 20554-84-1 Molecular Formula: C15H20O3 Molecular Weight (g/mol): 248.32 InChI Key: KTEXNACQROZXEV-QLIGOWBFSA-N IUPAC Name: (1S,2S,4R,7Z,11S)-4,8-dimethyl-12-methylidene-3,14-dioxatricyclo[9.3.0.0²,⁴]tetradec-7-en-13-one SMILES: C\C1=C\CC[C@@]2(C)O[C@H]2[C@H]2OC(=O)C(=C)[C@@H]2CC1
2
Parthenolide, MedChemExpress
MedChemExpress Parthenolide is a sesquiterpene lactone found in the medicinal herb Feverfew. Parthenolide exhibits anti-inflammatory activity by inhibiting NF-κB activation; also inhibits HDAC1 protein without affecting other class I/II HDACs.