missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals PARP Inhibitor VIII, PJ34

Product Code. 15456369
Change view
Click to view available options
Quantity:
5 mg
25 mg
Unit Size:
25mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15456369 5 mg 5mg
15466369 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15456369 Supplier Thermo Scientific Chemicals Supplier No. J64413.#0
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

A cell-permeable, water-soluble phenanthridinone-derivative that acts as a potent inhibitor of poly(ADP-ribose) polymerase

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • A cell-permeable, water-soluble phenanthridinone-derivative that acts as a potent inhibitor of poly(ADP-ribose) polymerase
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 344458-15-7
Molecular Formula C17H17N3O2·HCl, C17H18ClN3O2
Synonym 2-Dimethylamino-N-(6-oxo-5,6-dihydrophenanthridin-2-yl)acetamide hydrochloride

Specifications

Quantity 5 mg
Description White
Content And Storage -30°C to -10°C
For Use With (Application) A cell-permeable, water-soluble phenanthridinone-derivative that acts as a potent inhibitor of poly(ADP-ribose) polymerase
Formula Weight 331.80g/mol
Sensitivity Light Sensitive and Hygroscopic
Product Type Signal Transduction Reagent
Form Solid
InChI Key RURAZZMDMNRXMI-UHFFFAOYSA-N
SMILES Cl.CN(C)CC(=O)NC1=CC=C2NC(=O)C3=CC=CC=C3C2=C1
IUPAC Name 2-(dimethylamino)-N-(6-oxo-5,6-dihydrophenanthridin-2-yl)acetamide hydrochloride
Molecular Weight (g/mol) 331.80
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.