missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Parkin Antibody (PRK8), Novus Biologicals™
Mouse Monoclonal Antibody
£214.00 - £744.00
Specifications
| Antigen | Parkin |
|---|---|
| Clone | PRK8 |
| Dilution | Western Blot 1:100-1:2000, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunoprecipitation 1:10-1:500 |
| Applications | Immunohistochemistry, Immunofluorescence, Immunoprecipitation, Western Blot, Immunocytochemistry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18115804
|
Novus Biologicals
NBP2-29838-0.025ML |
0.025 mL |
£214.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18025374
|
Novus Biologicals
NBP2-29838 |
0.1 mL |
£744.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Parkin Monoclonal specifically detects Parkin in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation.Specifications
| Parkin | |
| Western Blot 1:100-1:2000, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunoprecipitation 1:10-1:500 | |
| Monoclonal | |
| Ascites | |
| RUO | |
| Human, Mouse | |
| O60260 | |
| 5071 | |
| Full length recombinant human Parkin. Epitope has been mapped to the second ring domain (aa 399-465). | |
| Primary | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
| PRK8 | |
| Immunohistochemistry, Immunofluorescence, Immunoprecipitation, Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Mouse | |
| Alzheimers Research, Neurodegeneration, Neuronal Cell Markers, Neuroscience, Phospho Specific | |
| Ascites, containing 30% glycerol. with 0.05% Sodium Azide | |
| E3 ubiquitin ligase, E3 ubiquitin-protein ligase parkin, EC 6.3.2.-, LPRS2, parkin, parkin 2, Parkinson disease protein 2, Parkinson juvenile disease protein 2, parkinson protein 2, E3 ubiquitin protein ligase (parkin), PDJjuvenile) 2, parkin | |
| PRKN | |
| IgG2b | |
| Unpurified | |
| Parkin 52kDa.May be observed as a doublet at 52 and 46kDa weight may be detected. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title