missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Parkin Antibody (PRK8), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-29838
This item is not returnable.
View return policy
Description
Parkin Monoclonal specifically detects Parkin in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation.
Specifications
| Parkin | |
| Monoclonal | |
| Unconjugated | |
| Ascites, containing 30% glycerol. with 0.05% Sodium Azide | |
| E3 ubiquitin ligase, E3 ubiquitin-protein ligase parkin, EC 6.3.2.-, LPRS2, parkin, parkin 2, Parkinson disease protein 2, Parkinson juvenile disease protein 2, parkinson protein 2, E3 ubiquitin protein ligase (parkin), PDJjuvenile) 2, parkin | |
| Mouse | |
| Unpurified | |
| RUO | |
| Primary | |
| Parkin 52kDa.May be observed as a doublet at 52 and 46kDa weight may be detected. | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. | |
| IgG2b |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation | |
| PRK8 | |
| Western Blot 1:100-1:2000, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunoprecipitation 1:10-1:500 | |
| O60260 | |
| PRKN | |
| Full length recombinant human Parkin. Epitope has been mapped to the second ring domain (aa 399-465). | |
| 0.1 mL | |
| Alzheimers Research, Neurodegeneration, Neuronal Cell Markers, Neuroscience, Phospho Specific | |
| 5071 | |
| Human, Mouse | |
| Ascites |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction