Learn More
PARG Mouse anti-Human, Unconjugated, Clone: 1H10G10, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 67946-1-IG-20UL
This item is not returnable.
View return policy
Description
The PARG antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human PARG. This antibody recognizes human antigen. The PARG antibody has been validated for the following applications: IHC, WB, ELISA analysis.
Specifications
| PARG | |
| Monoclonal | |
| Unconjugated | |
| 20 μL | |
| 8505 | |
| IgG2b |
| Western Blot, Immunohistochemistry | |
| 1H10G10 | |
| Mouse | |
| Primary | |
| Human |