missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PARG Mouse anti-Human, Unconjugated, Clone: 1H10G10, Proteintech
Mouse Monoclonal Antibody
£135.00 - £349.00
Specifications
| Antigen | PARG |
|---|---|
| Clone | 1H10G10 |
| Applications | Western Blot, Immunohistochemistry |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
17442443
|
Proteintech
67946-1-IG-20UL |
20 μL |
£135.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
17432443
|
Proteintech
67946-1-IG-150UL |
150 μL |
£349.00
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The PARG antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human PARG. This antibody recognizes human antigen. The PARG antibody has been validated for the following applications: IHC, WB, ELISA analysis.
Specifications
| PARG | |
| Western Blot, Immunohistochemistry | |
| Unconjugated | |
| Human | |
| IgG2b |
| 1H10G10 | |
| Monoclonal | |
| Mouse | |
| 8505 | |
| Primary |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title