Learn More
PARD3 Rabbit, anti-Human, CoraLite Plus 488, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
£135.00 - £375.00
Specifications
| Antigen | PARD3 |
|---|---|
| Concentration | 1000 μg/mL |
| Applications | Immunofluorescence |
| Conjugate | CoraLite Plus 488 |
| Form | Liquid |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18099795
|
Proteintech
CL488-11085-100UL |
100 μL |
£375.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18004755
|
Proteintech
CL488-11085-20UL |
20 μL |
£135.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The PARD3 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human PARD3. This antibody recognizes human, mouse, rat antigen. The PARD3 antibody has been validated for the following applications: IF analysis. This antibody is coralite™ plus 488.
Specifications
| PARD3 | |
| Immunofluorescence | |
| Liquid | |
| Human, Mouse, Rat | |
| ASIP, Baz, Bazooka, CTCL tumor antigen se2 5, FLJ21015, PAR 3, PAR3, PAR3 alpha, PAR3A, PAR3alpha, PARD 3, PARD3, PARD3A, SE2 5L16, SE2 5LT1, SE2 5T2 | |
| Primary |
| 1000 μg/mL | |
| CoraLite Plus 488 | |
| Rabbit | |
| 56288 | |
| IgG | |
| Store at -20°C. Avoid exposure to light. Stable for one year after shipment. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.