Learn More
PARD3 Rabbit, anti-Human, CoraLite Plus 488, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech CL488-11085-100UL
Description
The PARD3 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human PARD3. This antibody recognizes human, mouse, rat antigen. The PARD3 antibody has been validated for the following applications: IF analysis. This antibody is coralite™ plus 488.
Specifications
| PARD3 | |
| 1000 μg/mL | |
| ASIP, Baz, Bazooka, CTCL tumor antigen se2 5, FLJ21015, PAR 3, PAR3, PAR3 alpha, PAR3A, PAR3alpha, PARD 3, PARD3, PARD3A, SE2 5L16, SE2 5LT1, SE2 5T2 | |
| 100 μL | |
| 56288 | |
| Store at -20°C. Avoid exposure to light. Stable for one year after shipment. | |
| Liquid |
| Immunofluorescence | |
| CoraLite Plus 488 | |
| Rabbit | |
| Primary | |
| Human, Mouse, Rat | |
| 150 kDa | |
| IgG |