Learn More
Invitrogen™ PARD3 Monoclonal Antibody (K49032_10A1), Invitrogen™
Mouse Monoclonal Antibody
£380.00 - £570.00
Specifications
| Antigen | PARD3 |
|---|---|
| Clone | K49032_10A1 |
| Concentration | 1 mg/mL |
| Applications | Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30207492
|
Invitrogen™
MA555767 |
50 μL |
£380.00
50µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30204055
|
Invitrogen™
MA556147 |
200 μL |
£570.00
200µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Sequence of this protein is as follows: NQEKGDKTDR KKDKTGKEKK KDRDKEKDKM KAKKGMLKGL GDMFRFGKHR KDDKIEKTGK IKIQESFTSE EERIRMKQEQ ERIQAKTREF RERQARERDY AEIQDFHRTF GCDDELMYGG VSSYEGSMAL NARPQSPREG HMMDALYAQV KKPRNSKPSP VDSNRSTPSN HDRIQRLRQE FQQAKQDEDV EDRRRTYSFE QPWPNARPAT QSGRHSVSVE VQMQRQRQEE RESSQQAQRQ YSSLPRQSRK NASSVSQDSW EQNYSPGEGF QSAKENPRYS SYQGSRNGYL GGHGFNARVM LETQELLRQE QRRKEQQMKK QPPSEGPSNY DSYKKVQDPS YAPPKGPFRQ DVPPSPSQVA
PARD3 Monoclonal antibody specifically detects PARD3 in Human samples. It is validated for Western BlotSpecifications
| PARD3 | |
| 1 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol, 0.5% BSA and 0.09% sodium azide | |
| AA960621; AI256638; ASBP; ASIP; atypical PKC isotype-specific interacting protein; atypical PKC isotype-specific-interacting protein; atypical PKC-specific binding protein; atypical PKC-specific-binding protein; Baz; bazooka; CTCL tumor antigen se2-5; D8Ertd580e; Ephrin-interacting protein; FLJ21015; PAR3; Par-3; par-3 (partitioning defective 3) homolog; par-3 family cell polarity regulator; par-3 family cell polarity regulator alpha; par-3 partitioning defective 3 homolog; PAR3A; PAR3alpha; PAR3-alpha; Pard3; PARD-3; PARD3A; partitioning defective 3 homolog; partitioning-defective 3 homolog; PHIP; PPP1R118; protein phosphatase 1, regulatory subunit 118; RP11-406D17.1; SE2-5L16; SE2-5LT1; SE2-5T2; three-PDZ containing protein similar to C. elegans PAR3 (partitioning defect) | |
| Recombinant human PARD3 (immunogen range is 981-1340). | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Pard3 |
| K49032_10A1 | |
| Western Blot | |
| Unconjugated | |
| Mouse | |
| Human | |
| Q8TEW0 | |
| 56288 | |
| IgG1 | |
| Protein A | |
| Antibody |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.