Learn More
Invitrogen™ PARD3 Monoclonal Antibody (K49032_10A1), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen™ MA555767
Description
Sequence of this protein is as follows: NQEKGDKTDR KKDKTGKEKK KDRDKEKDKM KAKKGMLKGL GDMFRFGKHR KDDKIEKTGK IKIQESFTSE EERIRMKQEQ ERIQAKTREF RERQARERDY AEIQDFHRTF GCDDELMYGG VSSYEGSMAL NARPQSPREG HMMDALYAQV KKPRNSKPSP VDSNRSTPSN HDRIQRLRQE FQQAKQDEDV EDRRRTYSFE QPWPNARPAT QSGRHSVSVE VQMQRQRQEE RESSQQAQRQ YSSLPRQSRK NASSVSQDSW EQNYSPGEGF QSAKENPRYS SYQGSRNGYL GGHGFNARVM LETQELLRQE QRRKEQQMKK QPPSEGPSNY DSYKKVQDPS YAPPKGPFRQ DVPPSPSQVA
This gene encodes a member of the kinesin 4 or subfamily of kinesin related proteins. This subfamily is also referred to as the chromokinesins. The encoded protein is an ATP dependent microtubule-based motor protein that is involved in the intracellular transport of membranous organelles. This protein also associates with condensed chromosome arms and may be involved maintaining chromosome integrity during mitosis. This protein may also be involved in the organization of the central spindle prior to cytokinesis. A pseudogene of this gene is found on chromosome X.
Specifications
| PARD3 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 50% glycerol, 0.5% BSA and 0.09% sodium azide | |
| Q8TEW0 | |
| Pard3 | |
| Recombinant human PARD3 (immunogen range is 981-1340). | |
| 50 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Western Blot | |
| K49032_10A1 | |
| Unconjugated | |
| Pard3 | |
| AA960621; AI256638; ASBP; ASIP; atypical PKC isotype-specific interacting protein; atypical PKC isotype-specific-interacting protein; atypical PKC-specific binding protein; atypical PKC-specific-binding protein; Baz; bazooka; CTCL tumor antigen se2-5; D8Ertd580e; Ephrin-interacting protein; FLJ21015; PAR3; Par-3; par-3 (partitioning defective 3) homolog; par-3 family cell polarity regulator; par-3 family cell polarity regulator alpha; par-3 partitioning defective 3 homolog; PAR3A; PAR3alpha; PAR3-alpha; Pard3; PARD-3; PARD3A; partitioning defective 3 homolog; partitioning-defective 3 homolog; PHIP; PPP1R118; protein phosphatase 1, regulatory subunit 118; RP11-406D17.1; SE2-5L16; SE2-5LT1; SE2-5T2; three-PDZ containing protein similar to C. elegans PAR3 (partitioning defect) | |
| Mouse | |
| Protein A | |
| RUO | |
| 56288 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.