missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PAR2 Rabbit anti-Human, Mouse, Rat, Clone: 9X8E6, Novus Biologicals™
Rabbit Monoclonal Antibody
£167.00 - £415.00
Specifications
| Antigen | PAR2 |
|---|---|
| Clone | 9X8E6 |
| Dilution | Western Blot 1:500 - 1:2000, Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin |
| Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18373684
|
Novus Biologicals
NBP3-16555-20UL |
20 μg |
£167.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18311536
|
Novus Biologicals
NBP3-16555-100UL |
100 μg |
£415.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
PAR2 Monoclonal antibody specifically detects PAR2 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
| PAR2 | |
| Western Blot 1:500 - 1:2000, Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| coagulation factor II (thrombin) receptor-like 1, Coagulation factor II receptor-like 1, GPR11protease-activated receptor 2, G-protein coupled receptor 11, PAR-2, PAR2G protein-coupled receptor-11, proteinase-activated receptor 2, proteinase-activated receptor-2, Thrombin receptor-like 1 | |
| A synthetic peptide corresponding to a sequence within amino acids 298-397 of human PAR2 (P55085). ICFTPSNLLLVVHYFLIKSQGQSHVYALYIVALCLSTLNSCIDPFVYYFVSHDFRDHAKNALLCRSVRTVKQMQVSLTSKKHSRKSSSYSSSSTTVKTSY | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
| 9X8E6 | |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| GPCR, Signal Transduction | |
| PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
| 2150 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title