missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PAPOLG Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-55534-25ul
This item is not returnable.
View return policy
Description
PAPOLG Polyclonal specifically detects PAPOLG in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.
Specifications
| Elk1 (p Ser389) | |
| Polyclonal | |
| Unconjugated | |
| Antibody supplied in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, and 50% glycerol. with 0.02% Sodium Azide | |
| ELK1 | |
| Elk1 (Phospho-Ser389) antibody was raised against a peptide sequence around phosphorylation site of serine 389 (P-R-S (p) -P-A) derived from Human Elk-1. | |
| Epitope affinity purified | |
| RUO | |
| 2002 | |
| Human | |
| IgG |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| 1 mg/mL | |
| Western Blot 1:500-1:2000, Immunohistochemistry 1:50-1:100, Immunohistochemistry-Paraffin 1:10-1:500 | |
| ELK1, member of ETS oncogene family, ETS domain-containing protein Elk-1, ETS-like gene 1 | |
| Rabbit | |
| 62 kDa | |
| 0.025 mg | |
| Primary | |
| This antibody detects endogenous level of Elk-1 only when phosphorylated at serine 389. | |
| Store at -20°C. Avoid freeze/thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction