missing translation for 'onlineSavingsMsg'
Learn More

Papaveroxine, TRC

Product Code. 30416124
Change view
Click to view available options
Quantity:
10 mg
100 mg
Unit Size:
10mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30416124 10 mg 10mg
30409910 100 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30416124 Supplier Toronto Research Chemicals Supplier No. TRCP19052010MG
 Restricted Item
Estimated Shipment: 13-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 106982-92-7
Molecular Formula C22H25NO7
Molecular Weight (g/mol) 415.44
Synonym 6-[(S)-Hydroxy[(5R)-5,6,7,8-tetrahydro-4-methoxy-6-methyl-1,3-dioxolo[4,5-g]isoquinolin-5-yl]methyl]-2,3-dimethoxybenzaldehyde, [S-(R*,S*)]-6-[hydroxy(5,6,7,8-tetrahydro-4-methoxy-6-methyl-1,3-dioxolo[4,5-g]isoquinolin-5-yl)methyl]-2,3-dimethoxybenzaldehyde
IUPAC Name 6-[(S)-hydroxy-[(5R)-4-methoxy-6-methyl-7,8-dihydro-5H-[1,3]dioxolo[4,5-g]isoquinolin-5-yl]methyl]-2,3-dimethoxybenzaldehyde
SMILES COc1ccc([C@H](O)[C@@H]2N(C)CCc3cc4OCOc4c(OC)c23)c(C=O)c1OC

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C22H25NO7/c1-23-8-7-12-9-16-21(30-11-29-16)22(28-4)17(12)18(23)19(25)13-5-6-15(26-2)20(27-3)14(13)10-24/h5-6,9-10,18-19,25H,7-8,11H2,1-4H3/t18-,19+/m1/s1
Formula Weight 415.1631
Chemical Name or Material Papaveroxine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.