CAS RN 106982-92-7
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Papaveroxine, TRC
CAS: 106982-92-7 Molecular Formula: C22H25NO7 Molecular Weight (g/mol): 415.44 Synonym: 6-[(S)-Hydroxy[(5R)-5,6,7,8-tetrahydro-4-methoxy-6-methyl-1,3-dioxolo[4,5-g]isoquinolin-5-yl]methyl]-2,3-dimethoxybenzaldehyde,[S-(R*,S*)]-6-[hydroxy(5,6,7,8-tetrahydro-4-methoxy-6-methyl-1,3-dioxolo[4,5-g]isoquinolin-5-yl)methyl]-2,3-dimethoxybenzaldehyde IUPAC Name: 6-[(S)-hydroxy-[(5R)-4-methoxy-6-methyl-7,8-dihydro-5H-[1,3]dioxolo[4,5-g]isoquinolin-5-yl]methyl]-2,3-dimethoxybenzaldehyde SMILES: COc1ccc([C@H](O)[C@@H]2N(C)CCc3cc4OCOc4c(OC)c23)c(C=O)c1OC