missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals Papain, Carica Papaya Latex
Click to view available options
Quantity:
25 mg
100 mg
1 g
Beschreibung
Papain, from papaya latex, has been used to digest carotid artery smooth muscle cells. Papain is used in dissecting solutions. It is used to produce Fab fragments of antibodies. It is used for cell dissociation since it has been shown to be more effective and less damaging with certain tissues.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Spezifikation
Description | White to Off-white |
For Use With (Application) | A cysteine protease; breaks down intercellular matrix of cartilage |
Molecular Weight (g/mol) | 23 kd |
Content And Storage | 2°C to 8°C |
Form | Lyophilized powder,23 kDa |
Quantity | 1 g |
Product Type | Protease |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'