Learn More
STEMCELL Technologies Papain
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
Description
- Papain is a cysteine protease consisting of a single polypeptide containing three disulfide bridges
- Native crystalline papain is unreactive until acted upon by mild reducing agents, e.g. , cysteine, sulfide, or sulfite, and therefore likely exists as a zymogen
- Papain has a wide specificity with a preference towards arginine, lysine, and phenylalanine
- This enzyme degrades protein substrates such as the intercellular matrices of cartilage more extensively than pancreatic proteases and is typically less damaging and more effective than other proteases for tissue dissociation applications (Huettner & Baugham; Lam) and has also been used for the digestion of neural tissue (Fasano et al.)
Specifications
Specifications
| Description | Dopaminergic Neurons; Mesenchymal Cells, PSC-Derived; Neurons; Pancreatic Cells |
| Quantity | 25 mg |
| Synonym | Papainase; Papaya peptidase I |