missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pancreatic trypsin inhibitor Antibody (18) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
£156.00 - £398.00
Specifications
| Antigen | Pancreatic trypsin inhibitor |
|---|---|
| Clone | 18 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:100-1:2000, ELISA 1:100-1:2000 |
| Applications | Western Blot, ELISA |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18137002
|
Novus Biologicals
NBP2-23588-0.025ML |
0.025 mL |
£156.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18183525
|
Novus Biologicals
NBP2-23588-0.1ML |
0.1 mL |
£398.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Pancreatic trypsin inhibitor Monoclonal specifically detects Pancreatic trypsin inhibitor in Bovine samples. It is validated for Western Blot, ELISA.Specifications
| Pancreatic trypsin inhibitor | |
| 1 mg/mL | |
| Western Blot, ELISA | |
| Unconjugated | |
| Mouse | |
| 10mM Phosphate (pH 7.4) and 0.5M NaCl with 15mM Sodium Azide | |
| 404172 | |
| IgG1 κ | |
| Protein A or G purified | |
| Clone 18 binds bovine aprotinin immobilized on a solid phase or free in solution. |
| 18 | |
| Western Blot 1:100-1:2000, ELISA 1:100-1:2000 | |
| Monoclonal | |
| Purified | |
| Bovine | |
| Aprotinin, Basic protease inhibitor, BPI, BPTI, PTI, PTI pancreatic trypsin inhibitor | |
| Aprotinin from bovine lung adsorbed onto aluminum hydroxide gel | |
| Primary | |
| Store at 4C. Do not freeze. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title