missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pancreatic trypsin inhibitor Antibody (18) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-23588-0.1ML
This item is not returnable.
View return policy
Description
Pancreatic trypsin inhibitor Monoclonal specifically detects Pancreatic trypsin inhibitor in Bovine samples. It is validated for Western Blot, ELISA.
Specifications
| Pancreatic trypsin inhibitor | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:100-1:2000, ELISA 1:100-1:2000 | |
| Aprotinin, Basic protease inhibitor, BPI, BPTI, PTI, PTI pancreatic trypsin inhibitor | |
| Aprotinin from bovine lung adsorbed onto aluminum hydroxide gel | |
| 0.1 mL | |
| 404172 | |
| Bovine | |
| Purified |
| Western Blot, ELISA | |
| 18 | |
| Unconjugated | |
| 10mM Phosphate (pH 7.4) and 0.5M NaCl with 15mM Sodium Azide | |
| Mouse | |
| Protein A or G purified | |
| Primary | |
| Clone 18 binds bovine aprotinin immobilized on a solid phase or free in solution. | |
| Store at 4C. Do not freeze. | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction