missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PAK4 Recombinant Human Protein
PAK4 is a serine/ threonine p21 activating kinase.
Brand: Invitrogen™ PV4284
Code : NEW
Additional Details : Weight : 0.01000kg
Description
PAK4 is implicated in the reorganization of actin cytoskeleton and in the formation of filopodia.
Specifications
NP_005875 | |
50mM Tris pH 7.5, 150mM NaCl, 0.5mM EDTA, 0.02% Triton X-100, 2mM DTT, 50% Glycerol | |
1mg | |
RUO - research use only | |
PAK4 | |
Human | |
GST Tag | |
Frozen |
Enzyme and Protein Activity Assay, Kinase Assay, Protein Assays and Analysis, Protein Biology | |
61.6kDa | |
Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws. | |
Catalytic | |
STE20 Kinase Family | |
Recombinant | |
Baculovirus |