missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PAK4 Recombinant Human Protein
PAK4 is a serine/ threonine p21 activating kinase.
£1602.00 - £6900.00
Specifications
Accession Number | NP_005875 |
---|---|
For Use With (Application) | Enzyme and Protein Activity Assay, Kinase Assay, Protein Assays and Analysis, Protein Biology |
Formulation | 50mM Tris pH 7.5, 150mM NaCl, 0.5mM EDTA, 0.02% Triton X-100, 2mM DTT, 50% Glycerol |
Molecular Weight (g/mol) | 61.6kDa |
Storage Requirements | Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15438633
|
Invitrogen™
PR7856A |
100μg |
£1602.00
100µg |
Estimated Shipment: 10-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15478723
|
Invitrogen™
PV4284 |
1mg |
£6900.00
1mg |
Estimated Shipment: 17-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
PAK4 is implicated in the reorganization of actin cytoskeleton and in the formation of filopodia.
Specifications
NP_005875 | |
50mM Tris pH 7.5, 150mM NaCl, 0.5mM EDTA, 0.02% Triton X-100, 2mM DTT, 50% Glycerol | |
Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws. | |
Catalytic | |
STE20 Kinase Family | |
Recombinant | |
Baculovirus |
Enzyme and Protein Activity Assay, Kinase Assay, Protein Assays and Analysis, Protein Biology | |
61.6kDa | |
RUO - research use only | |
PAK4 | |
Human | |
GST Tag | |
Frozen |