missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PAK4 Monoclonal Antibody (K1E023_12G11), Invitrogen™
Mouse Monoclonal Antibody
£453.00 - £760.00
Specifications
| Antigen | PAK4 |
|---|---|
| Clone | K1E023_12G11 |
| Concentration | 1 mg/mL |
| Applications | Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30209369
|
Invitrogen™
MA556314 |
200 μL |
£760.00
200µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30204447
|
Invitrogen™
MA555934 |
50 μL |
£453.00
50µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Sequence of this protein is as follows: RRDSPPPPAR ARQENGMPEE PATTARGGPG KAGSRGRFAG HSEAGGGSGD RRRAGPEKRP KSSREGSGGP QESSRDKRPL SGPDVGTPQP AGLASGAKLA AGRPFNTYPR ADTDHPSRGA
PAK4 Monoclonal antibody specifically detects PAK4 in Human samples. It is validated for Immunohistochemistry (Paraffin), Immunoprecipitation, Western BlotSpecifications
| PAK4 | |
| 1 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol, 0.5% BSA and 0.09% sodium azide | |
| 5730488L07Rik; AW555722; hypothetical protein LOC431769; im:5916170; Kiaa1142; mKIAA1142; p21 (CDKN1A)-activated kinase 4; p21 (RAC1) activated kinase 4; p21 protein (Cdc42/Rac)-activated kinase 4; p21(CDKN1A)-activated kinase 4; p21-activated kinase 4; Pak4; PAK-4; protein kinase related to S. cerevisiae STE20, effector for Cdc42Hs; serine/threonine-protein kinase PAK 4; wu:fd12b01; zgc:92014 | |
| Recombinant human PAK4 (immunogen range is 101-220). | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Pak4 |
| K1E023_12G11 | |
| Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| Unconjugated | |
| Mouse | |
| Human | |
| O96013 | |
| 10298 | |
| IgG1 | |
| Protein A | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title