missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PAIP1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-68963-25ul
This item is not returnable.
View return policy
Description
PAIP1 Polyclonal antibody specifically detects PAIP1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| PAIP1 | |
| Polyclonal | |
| Western Blot 0.04-0.4 μg/ml, Immunohistochemistry 1:1000 - 1:2500, Immunohistochemistry-Paraffin 1:1000 - 1:2500 | |
| MGC12360, PABC1-interacting protein 1, PABP-interacting protein 1, PAIP-1, poly(A) binding protein interacting protein 1, Poly(A)-binding protein-interacting protein 1, polyadenylate binding protein-interacting protein 1, polyadenylate-binding protein-interacting protein 1 | |
| This antibody was developed against a recombinant protein corresponding to amino acids: GTNGQVTRADILQVGLRELLNALFSNPMDDNLICAVKLLKLTGSVLEDAWKEKGKMDMEEIIQRIENVVLDANCSRDVKQMLLK | |
| 25 μL | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.2) and 40% Glycerol | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| 10605 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction