missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PAGE1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-58313
This item is not returnable.
View return policy
Description
PAGE1 Polyclonal specifically detects PAGE1 in Human samples. It is validated for Western Blot.
Specifications
| PAGE1 | |
| Polyclonal | |
| Unconjugated | |
| PBS, 2% Sucrose with 0.09% Sodium Azide | |
| CT16.3, G antigen, family B, 1 (prostate associated), GAGE9, GAGE-9AL5, GAGEB1G antigen family B member 1, P antigen family, member 1 (prostate associated), PAGE-1G antigen 9, prostate associated gene 1, Prostate-associated gene 1 protein | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| 8712 | |
| Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 100μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| 1 mg/ml | |
| Western Blot 1.0 ug/ml | |
| O75459 | |
| PAGE1 | |
| Synthetic peptides corresponding to PAGE1(P antigen family, member 1 (prostate associated)) The peptide sequence was selected from the middle region of PAGE1. Peptide sequence TKRVCLRNEEQMKLPAEGPEPEADSQEQVHPKTGCERGDGPDVQELGLPN. | |
| 100 μL | |
| Primary | |
| Expected identity based on immunogen sequence: Human: 100%;. | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction