missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PAG3 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£157.00 - £301.00
Specifications
| Antigen | PAG3 |
|---|---|
| Concentration | 0.5 mg/mL |
| Dilution | Western Blot 1.0-2.0 ug/ml |
| Applications | Western Blot |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18043524
|
Novus Biologicals
NBP2-24614SS |
0.025 mg |
£157.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18766583
|
Novus Biologicals
NBP2-24614 |
£301.00
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||||
Description
PAG3 Polyclonal specifically detects PAG3 in Human, Mouse, Rat, Opossum, Primate samples. It is validated for Western Blot.Specifications
| PAG3 | |
| Western Blot 1.0-2.0 ug/ml | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat, Opossum, Primate | |
| O43150 | |
| 8853 | |
| A portion of amino acids 700-750 of human ASAP2 was used as the immunogen. | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| 0.5 mg/mL | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Cellular Markers, Cytoskeleton Markers | |
| 0.2 ml PBS and 0.05% BSA with 0.05% Sodium Azide | |
| arf-GAP with SH3 domain, ANK repeat and PH domain-containing protein 2, ArfGAP with SH3 domain, ankyrin repeat and PH domain 2, centaurin, beta 3, CENTB3, DDEF2FLJ42910, development and differentiation enhancing factor 2, Development and differentiation-enhancing factor 2, KIAA0400AMAP2, Pap-alpha, PAPPAG3, Paxillin-associated protein with ARF GAP activity 3, PYK2 C terminus-associated protein, Pyk2 C-terminus-associated protein, SHAG1 | |
| ASAP2 | |
| IgG | |
| Protein A purified | |
| The amino acid sequence used as immunogen is 100% homologous in human (isoforms CRA_a, CRA_c and CRA_e), chimpanzee and monkey, 86% homologous in horse, 83% homologous in mouse, rat, and cow, and 73% homologous in dog. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title