missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PAG3 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-24614SS
This item is not returnable.
View return policy
Description
PAG3 Polyclonal specifically detects PAG3 in Human, Mouse, Rat, Opossum, Primate samples. It is validated for Western Blot.
Specifications
| PAG3 | |
| Polyclonal | |
| Unconjugated | |
| 0.2 ml PBS and 0.05% BSA with 0.05% Sodium Azide | |
| arf-GAP with SH3 domain, ANK repeat and PH domain-containing protein 2, ArfGAP with SH3 domain, ankyrin repeat and PH domain 2, centaurin, beta 3, CENTB3, DDEF2FLJ42910, development and differentiation enhancing factor 2, Development and differentiation-enhancing factor 2, KIAA0400AMAP2, Pap-alpha, PAPPAG3, Paxillin-associated protein with ARF GAP activity 3, PYK2 C terminus-associated protein, Pyk2 C-terminus-associated protein, SHAG1 | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| The amino acid sequence used as immunogen is 100% homologous in human (isoforms CRA_a, CRA_c and CRA_e), chimpanzee and monkey, 86% homologous in horse, 83% homologous in mouse, rat, and cow, and 73% homologous in dog. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| 0.5 mg/mL | |
| Western Blot 1.0-2.0 ug/ml | |
| O43150 | |
| ASAP2 | |
| A portion of amino acids 700-750 of human ASAP2 was used as the immunogen. | |
| 0.025 mg | |
| Cellular Markers, Cytoskeleton Markers | |
| 8853 | |
| Human, Mouse, Rat, Opossum, Primate | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction