missing translation for 'onlineSavingsMsg'
Läs mer
Läs mer
PAG3 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
£151.00 - £366.00
Specifikationer
| Antigen | PAG3 |
|---|---|
| Dilution | Western Blot 1:500-1:2000 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Produktkod | Brand | Quantity | Pris | Kvantitet och tillgänglighet | |||||
|---|---|---|---|---|---|---|---|---|---|
| Produktkod | Brand | Quantity | Pris | Kvantitet och tillgänglighet | |||||
|
18648971
|
Novus Biologicals
NBP2-94260-0.02ml |
0.02 mL |
£151.00
0.02mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18690212
|
Novus Biologicals
NBP2-94260-0.1ml |
0.1 mL |
£366.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Beskrivning
PAG3 Polyclonal antibody specifically detects PAG3 in Mouse samples. It is validated for Western BlotSpecifikationer
| PAG3 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Cell Biology, Cellular Markers | |
| PBS (pH 7.3), 50% glycerol | |
| 8853 | |
| IgG | |
| Affinity purified |
| Western Blot 1:500-1:2000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Mouse | |
| arf-GAP with SH3 domain, ANK repeat and PH domain-containing protein 2, ArfGAP with SH3 domain, ankyrin repeat and PH domain 2, centaurin, beta 3, CENTB3, DDEF2FLJ42910, development and differentiation enhancing factor 2, Development and differentiation-enhancing factor 2, KIAA0400AMAP2, Pap-alpha, PAPPAG3, Paxillin-associated protein with ARF GAP activity 3, PYK2 C terminus-associated protein, Pyk2 C-terminus-associated protein, SHAG1 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-120 of human ASAP2 (NP_001128663). MPDQISVSEFVAETHEDYKAPTASSFTTRTAQCRNTVAAIEEALDVDRMVLYKMKKSVKAINSSGLAHVENEEQYTQALEKFGGNCVCRDDPDLGSAFLKFSVFTKELTALFKNLIQNMN | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel