missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PAG3 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-94260-0.1ml
This item is not returnable.
View return policy
Description
PAG3 Polyclonal antibody specifically detects PAG3 in Mouse samples. It is validated for Western Blot
Specifications
| PAG3 | |
| Polyclonal | |
| Western Blot 1:500-1:2000 | |
| arf-GAP with SH3 domain, ANK repeat and PH domain-containing protein 2, ArfGAP with SH3 domain, ankyrin repeat and PH domain 2, centaurin, beta 3, CENTB3, DDEF2FLJ42910, development and differentiation enhancing factor 2, Development and differentiation-enhancing factor 2, KIAA0400AMAP2, Pap-alpha, PAPPAG3, Paxillin-associated protein with ARF GAP activity 3, PYK2 C terminus-associated protein, Pyk2 C-terminus-associated protein, SHAG1 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-120 of human ASAP2 (NP_001128663). MPDQISVSEFVAETHEDYKAPTASSFTTRTAQCRNTVAAIEEALDVDRMVLYKMKKSVKAINSSGLAHVENEEQYTQALEKFGGNCVCRDDPDLGSAFLKFSVFTKELTALFKNLIQNMN | |
| 0.1 mL | |
| Cell Biology, Cellular Markers | |
| 8853 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction