missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PAFAH1B2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£362.00
Specifications
| Antigen | PAFAH1B2 |
|---|---|
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
Description
PAFAH1B2 Polyclonal specifically detects PAFAH1B2 in Human samples. It is validated for Western Blot.Specifications
| PAFAH1B2 | |
| Polyclonal | |
| Rabbit | |
| O35264 | |
| 5049 | |
| IgG | |
| This product is specific to Subunit or Isoform: beta. |
| Western Blot | |
| Unconjugated | |
| RUO | |
| EC 3.1.1.47, intracellular platelet-activating factor acetylhydrolase alpha 2 subunit, PAF acetylhydrolase 30 kDa subunit, PAF-AH 30 kDa subunit, PAFAH subunit beta, PAF-AH subunit beta, PAF-AH1b alpha 2 subunit, PAFAHB, platelet-activating factor acetylhydrolase 1b, catalytic subunit 2 (30kDa), platelet-activating factor acetylhydrolase IB subunit beta, platelet-activating factor acetylhydrolase, isoform Ib, beta subunit 30kDa, platelet-activating factor acetylhydrolase, isoform Ib, subunit 2 (30kDa) | |
| PAFAH1B2 | |
| Primary |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title