missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PAFAH1B2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-56301
This item is not returnable.
View return policy
Description
PAFAH1B2 Polyclonal specifically detects PAFAH1B2 in Human samples. It is validated for Western Blot.
Specifications
| PAFAH1B2 | |
| Polyclonal | |
| Unconjugated | |
| PBS, 2% Sucrose with 0.09% Sodium Azide | |
| EC 3.1.1.47, intracellular platelet-activating factor acetylhydrolase alpha 2 subunit, PAF acetylhydrolase 30 kDa subunit, PAF-AH 30 kDa subunit, PAFAH subunit beta, PAF-AH subunit beta, PAF-AH1b alpha 2 subunit, PAFAHB, platelet-activating factor acetylhydrolase 1b, catalytic subunit 2 (30kDa), platelet-activating factor acetylhydrolase IB subunit beta, platelet-activating factor acetylhydrolase, isoform Ib, beta subunit 30kDa, platelet-activating factor acetylhydrolase, isoform Ib, subunit 2 (30kDa) | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 5049 | |
| Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| Western Blot | |
| 0.5 mg/ml | |
| Western Blot 1.0 ug/ml | |
| O35264 | |
| PAFAH1B2 | |
| Synthetic peptides corresponding to PAFAH1B2(platelet-activating factor acetylhydrolase, isoform Ib, beta subunit 30kDa) The peptide sequence was selected from the N terminal of PAFAH1B2. Peptide sequence MSQGDSNPAAIPHAAEDIQGDDRWMSQHNRFVLDCKDKEPDVLFVGDSMV The peptide sequence for this immunogen was taken from within the described region. | |
| 100 μL | |
| Primary | |
| This product is specific to Subunit or Isoform: beta. | |
| Human, Rat, Pig, Bovine, Canine, Equine, Rabbit | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido