missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PADI2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 12110-1-AP-150UL
This item is not returnable.
View return policy
Description
This gene encodes a member of the peptidyl arginine deiminase family of enzymes, which catalyze the post-translational deimination of proteins by converting arginine residues into citrullines in the presence of calcium ions. The family members have distinct substrate specificities and tissue-specific expression patterns. The type II enzyme is the most widely expressed family member. Known substrates for this enzyme include myelin basic protein in the central nervous system and vimentin in skeletal muscle and macrophages. This enzyme is thought to play a role in the onset and progression of neurodegenerative human disorders, including Alzheimer disease and multiple sclerosis, and it has also been implicated in glaucoma pathogenesis. This gene exists in a cluster with four other paralogous genes.Specifications
| PADI2 | |
| Polyclonal | |
| Unconjugated | |
| PADI2 | |
| KIAA0994, PAD H19, PAD2, PADI2, PDI2, Peptidylarginine deiminase II | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 11240, 18600, 29511 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| 0.22 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| P20717, Q08642, Q9Y2J8 | |
| Padi2 | |
| PADI2 Fusion Protein Ag2755 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction