missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Pacific Blue™ Succinimidyl Ester

Product Code. 11549196
Change view
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11549196 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 
Product Code. 11549196 Supplier Invitrogen™ Supplier No. P10163
Curiosity Goes Beyond
Use promo code 27966
View all promotions
 Restricted Item
Estimated Shipment: 10-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Pacific Blue™ succinimidyl ester

The amine-reactive Pacific Blue™ succinimidyl ester can be used to can be used to create blue-fluorescent bioconjugates with excitation/emission maxima ∼410/455 nm that are excitable by the 405 nm spectral line of the blue diode (violet) laser.

View all Pacific Blue™ dye products..

View the Fluorophore Selection Guide.
TRUSTED_SUSTAINABILITY

Specifications

Format Solid
Quantity 5 mg
Product Type Succinimidyl Ester
Solubility DMSO (Dimethylsulfoxide)
Content And Storage Contains 1 vial (1 mg). Store in freezer (-5 to -30°C) and protect from light.
Chemical Reactivity Amine
Detection Method Fluorescence
Flow Cytometer Laser Lines 405
Shipping Condition Room Temperature
Reactive Moiety Active Ester, Succinimidyl Ester
Label or Dye Pacific Blue™
Label Type Pacific Dyes
Product Line Pacific Blue
Emission 451
Excitation 416
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.