missing translation for 'onlineSavingsMsg'
Learn More

PA2G4 Mouse anti-Human, Mouse, Rat, Clone: 3G10D9, Proteintech

Mouse Monoclonal Antibody

Brand:  Proteintech 66055-1-IG-20UL

 View more versions of this product

Product Code. 16857585

  • £135.00 / 20µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

This antibody is a mouse mono-clonal antibody raised against full length PA2G4 of human origin.

This gene encodes an RNA-binding protein that is involved in growth regulation. This protein is present in pre-ribosomal ribonucleoprotein complexes and may be involved in ribosome assembly and the regulation of intermediate and late steps of rRNA processing. This protein can interact with the cytoplasmic domain of the ErbB3 receptor and may contribute to transducing growth regulatory signals. This protein is also a transcriptional co-repressor of androgen receptor-regulated genes and other cell cycle regulatory genes through its interactions with histone deacetylases. This protein has been implicated in growth inhibition and the induction of differentiation of human cancer cells. Six pseudogenes, located on chromosomes 3, 6, 9, 18, 20 and X, have been identified.
Specifications

Specifications

PA2G4
Monoclonal
1.04 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P50580, Q6AYD3, Q9UQ80
Pa2g4
PA2G4 Fusion Protein Ag7850
20 μL
Primary
Mouse, Human, Rat
Antibody
IgG2a
Immunohistochemistry (Paraffin), Immunofluorescence, Immunocytochemistry, Western Blot
3G10D9
Unconjugated
PA2G4
EBP1, ErbB3 binding protein 1, HG4 1, p38 2G4, PA2G4
Mouse
Protein A
RUO
18813, 288778, 5036
-20°C
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.