missing translation for 'onlineSavingsMsg'
Learn More

PA2G4 Mouse anti-Human, Mouse, Rat, Clone: 3G10D9, Proteintech

Mouse Monoclonal Antibody

£135.00 - £349.00

Specifications

Antigen PA2G4
Clone 3G10D9
Concentration 1.04 mg/mL
Applications Immunohistochemistry (Paraffin), Immunofluorescence, Immunocytochemistry, Western Blot
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16857585
View Documents
Proteintech
66055-1-IG-20UL
20 μL
£135.00
20µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
16847585
View Documents
Proteintech
66055-1-IG-150UL
150 μL
£349.00
150µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This antibody is a mouse mono-clonal antibody raised against full length PA2G4 of human origin.

This gene encodes an RNA-binding protein that is involved in growth regulation. This protein is present in pre-ribosomal ribonucleoprotein complexes and may be involved in ribosome assembly and the regulation of intermediate and late steps of rRNA processing. This protein can interact with the cytoplasmic domain of the ErbB3 receptor and may contribute to transducing growth regulatory signals. This protein is also a transcriptional co-repressor of androgen receptor-regulated genes and other cell cycle regulatory genes through its interactions with histone deacetylases. This protein has been implicated in growth inhibition and the induction of differentiation of human cancer cells. Six pseudogenes, located on chromosomes 3, 6, 9, 18, 20 and X, have been identified.
Specifications

Specifications

PA2G4
1.04 mg/mL
Monoclonal
Liquid
RUO
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
EBP1, ErbB3 binding protein 1, HG4 1, p38 2G4, PA2G4
Pa2g4
IgG2a
Protein A
Antibody
3G10D9
Immunohistochemistry (Paraffin), Immunofluorescence, Immunocytochemistry, Western Blot
Unconjugated
Mouse
Mouse, Human, Rat
P50580, Q6AYD3, Q9UQ80
18813, 288778, 5036
PA2G4 Fusion Protein Ag7850
Primary
-20°C
PA2G4
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.