missing translation for 'onlineSavingsMsg'
Learn More

p73α /β Ab-2, Mouse Monoclonal Antibody,Lab Vision™ Epredia™

Deliver accurate, reproducible results with the Epredia™ p73α/β Ab-2, Mouse Monoclonal Antibody.

Brand:  Epredia MS-762-P1ABX

 View more versions of this product

Product Code. 12682267

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

This item is not returnable. View return policy

Description

Description

p73 exhibits sequence homology to the p53's transcriptional activation, DNA binding, and oligomerization domains. However, adenovirus E1B 55K, simian virus 40 T, and human papillomavirus E6 do not physically interact with p73 indicating that the functions of these two proteins may differ under physiological conditions. p73 is expressed in either a full-length form (α) (MW ∽80kDa) or a shorter (Β) (MW ∽70kDa) mRNA variant, with exon 13 spliced out. Transient overexpression of p73 in cells can induce apoptosis and p21, a cellular p53 target gene primarily responsible for p53-dependent cell cycle arrest.

Host Species: Mouse

Clone: ER-15

Isotype: IgG1/κ

Species Reactivity: Human and Monkey. Others not known.

Epitope: aa 380-637 (p73α)

Immunogen: Purified monkey recombinant p73α protein (aa 380-637)

Molecular Weight: 80kDa (p73α) and 70kDa (p73β)

Positive Control: COS monkey kidney cells and IM9 human myeloma cells; SK-N-AS human neuroblastoma cells lack p73α/β

Cellular Localization: Nuclear Ab-2 binds to both p73α and p73β in immunoprecipitation and Western Blot assays. Circumstantial evidence suggestes that the epitopes of Ab-2 and Ab-3 are different.

Recommended for:

  • Western Blotting
  • Immunofluorescence
  • Immunoprecipitation
  • Gel Supershift

Note:

USA: RUO; Int'l: RUO

TRUSTED_SUSTAINABILITY
Specifications

Specifications

p73alpha/beta Ab-2
Monoclonal
1mg/mL
Mouse
100 μL
Cell Cycle
Human, Non-human Primate
IgG2a κ
Immunofluorescence, Western Blot
ER-15
Unconjugated
Purified monkey recombinant p73alpha protein (aa380-637)
RUO
Primary
Concentrated
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.