missing translation for 'onlineSavingsMsg'
Learn More
Learn More
p66 beta Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB100-56387SS
This item is not returnable.
View return policy
Description
p66 beta Polyclonal specifically detects p66 beta in Human, Mouse samples. It is validated for Western Blot.
Specifications
| p66 beta | |
| Polyclonal | |
| Unconjugated | |
| PBS containing 0.05% BSA with 0.05% Sodium Azide | |
| GATAD2B | |
| This antibody was developed against a synthetic peptide corresponding to aa 445-460 of mouse p66b. | |
| 0.025 mg | |
| Primary | |
| Human, Mouse | |
| Purified |
| Western Blot | |
| 0.5 mg/mL | |
| Western Blot 2-4 ug/ml | |
| FLJ37346, GATA zinc finger domain containing 2B, GATA zinc finger domain-containing protein 2B, KIAA1150, MGC138257, MGC138285, p66/p68, P66beta, RP11-216N14.6, transcription repressor p66 beta component of the MeCP1 complex, transcriptional repressor p66-beta | |
| Rabbit | |
| Protein G purified | |
| RUO | |
| 57459 | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction