missing translation for 'onlineSavingsMsg'
Learn More
Learn More
p66 beta Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£156.00 - £301.00
Specifications
| Antigen | p66 beta |
|---|---|
| Concentration | 0.5 mg/mL |
| Dilution | Western Blot 2-4 ug/ml |
| Applications | Western Blot |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18042714
|
Novus Biologicals
NB100-56387SS |
0.025 mg |
£156.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18778693
|
Novus Biologicals
NB100-56387 |
£301.00
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||||
Description
p66 beta Polyclonal specifically detects p66 beta in Human, Mouse samples. It is validated for Western Blot.Specifications
| p66 beta | |
| Western Blot 2-4 ug/ml | |
| Polyclonal | |
| Purified | |
| RUO | |
| PBS containing 0.05% BSA with 0.05% Sodium Azide | |
| 57459 | |
| This antibody was developed against a synthetic peptide corresponding to aa 445-460 of mouse p66b. | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| 0.5 mg/mL | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse | |
| FLJ37346, GATA zinc finger domain containing 2B, GATA zinc finger domain-containing protein 2B, KIAA1150, MGC138257, MGC138285, p66/p68, P66beta, RP11-216N14.6, transcription repressor p66 beta component of the MeCP1 complex, transcriptional repressor p66-beta | |
| GATAD2B | |
| IgG | |
| Protein G purified |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title