missing translation for 'onlineSavingsMsg'
Learn More

p63/TP73L Antibody (TP63/11), Novus Biologicals™

Mouse Monoclonal Antibody

£218.00 - £396.00

Specifications

Antigen p63/TP73L
Clone TP63/11
Concentration 0.2 mg/mL
Dilution Immunohistochemistry-Paraffin 0.25-0.5 μg/mL
Applications Immunohistochemistry (Paraffin)
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18633127
View Documents
Novus Biologicals
NBP3-07431-100ug
100 μg
£396.00
100µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18665186
View Documents
Novus Biologicals
NBP3-07431-20ug
20 μg
£218.00
20µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

p63/TP73L Monoclonal antibody specifically detects p63/TP73L in Human, Mouse samples. It is validated for Immunohistochemistry (Paraffin)
Specifications

Specifications

p63/TP73L
0.2 mg/mL
Immunohistochemistry (Paraffin)
Unconjugated
Mouse
Apoptosis, Cancer, Cellular Markers, Hypoxia, Stem Cell Markers, Tumor Suppressors
10 mM PBS with 0.05% BSA
amplified in squamous cell carcinoma, Chronic μgcerative stomatitis protein, CUSP, EEC3B(p51B), Keratinocyte transcription factor KET, KETRHS, LMS, OFC8SHFM4TP73LNBP, p40, p51tumor protein p53-like, p53CP, P63, p63AIS, p73H, P73L, p73LB(p51A), TP53CP, TP53L, Transformation-related protein 63, tumor protein 63, tumor protein p53-competing protein, tumor protein p63, tumor protein p63 deltaN isoform delta, Tumor protein p73-like
Recombinant full-length human p63/TP73L protein (Uniprot: Q9H3D4)
Primary
Store at 4°C.
TP63/11
Immunohistochemistry-Paraffin 0.25-0.5 μg/mL
Monoclonal
Purified
RUO
Human, Mouse
Q9H3D4, Q9H3D4
8626
IgG2a κ
Protein A or G purified
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.